This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00626862_g1
See other PLD4 GE Assays ›
Gene Symbol:
Pld4
Gene Name:
phospholipase D family, member 4
Gene Aliases:
AI132321, thss
Chromosome Location:
Chr.12: 112760655 - 112768988 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00626862_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pld4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178911.4 NP_849242.1 3 - 4 379 76
GenBank mRNA AK045459.1 - 3 - 4 338 76
AK079549.1 - 3 - 4 338 76
AK079878.1 - 3 - 4 381 76
AK151551.1 - 3 - 4 613 76
AK152969.1 - 2 - 3 226 76
AK170362.1 - 3 - 4 336 76
BC058565.1 - 3 - 4 329 6442038 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001272.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pld4

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
lipid metabolic process
phagocytosis
lipid catabolic process
catalytic activity
phospholipase D activity
protein binding
hydrolase activity
N-acylphosphatidylethanolamine-specific phospholipase D activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200009156.mm.1 NM_178911.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200009156.mm.1 NM_178911.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200009156.mm.1 TC1200001272.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0