This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00626868_g1
See other PLD4 GE Assays ›
Gene Symbol:
Pld4
Gene Name:
phospholipase D family, member 4
Gene Aliases:
AI132321, thss
Chromosome Location:
Chr.12: 112760655 - 112768988 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00626868_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pld4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178911.4 NP_849242.1 9 - 10 1315 54
GenBank mRNA AK045459.1 - 9 - 10 1274 54
AK079549.1 - 9 - 10 1274 54
AK079878.1 - 9 - 10 1317 54
AK151551.1 - 9 - 10 1549 54
AK152969.1 - 8 - 9 1164 54
AK170362.1 - 9 - 10 1272 54
BC058565.1 - 9 - 10 1265 6442038 54

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001272.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pld4

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
lipid metabolic process
phagocytosis
lipid catabolic process
catalytic activity
phospholipase D activity
protein binding
hydrolase activity
N-acylphosphatidylethanolamine-specific phospholipase D activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200009162.mm.1 NM_178911.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200009162.mm.1 NM_178911.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200009162.mm.1 TC1200001272.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0