This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00626973_m1
See other ADPRH GE Assays ›
Gene Symbol:
Adprh
Gene Name:
ADP-ribosylarginine hydrolase
Gene Aliases:
Arh1
Chromosome Location:
Chr.16: 38445399 - 38452689 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00626973_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adprh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007414.3 NP_031440.1 1 - 2 162 62
GenBank mRNA AF244347.1 - 131 62
AK010501.1 - 1 - 2 128 62
AK146280.1 - 1 - 2 132 62
AK153498.1 - 1 - 2 127 62
AK171798.1 - 1 - 2 132 62
BC003437.1 - 1 - 2 120 2647247 62
BC058232.1 - 1 - 2 120 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF244347.1, AK146280.1, AK153498.1, AK171798.1, BC003437.1, NM_007414.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adprh

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
protein de-ADP-ribosylation
magnesium ion binding
ADP-ribosylarginine hydrolase activity
hydrolase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600013127.mm.1 NM_007414.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600013127.mm.1 NM_007414.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600013127.mm.1 TC1600001605.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600013131.mm.1 NM_007414.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600013131.mm.1 NM_007414.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600013131.mm.1 TC1600001605.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600013132.mm.1 NM_007414.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600013132.mm.1 NM_007414.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600013132.mm.1 TC1600001605.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 906126 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline