This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00627597_m1
See other UCP2 GE Assays ›
Gene Symbol:
Ucp2
Gene Name:
uncoupling protein 2 (mitochondrial, proton carrier)
Gene Aliases:
Slc25a8
Chromosome Location:
Chr.7: 100493358 - 100499630 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00627597_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ucp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011671.5 NP_035801.3 1 - 2 106 69
GenBank mRNA AK035298.1 - 101 69
AK054060.1 - 107 69
AK134116.1 - 1 - 2 107 69
AK142687.1 - 1 - 2 107 69
AK165245.1 - 1 - 2 107 69
AK172307.1 - 1 - 2 69
BC012697.1 - 1 - 2 105 4205625 69
BY299894.1 - 1 - 2 129 69
U69135.1 - 1 - 2 34 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK035298.1, AK054060.1, AK134116.1, AK142687.1, BC012697.1, NM_011671.5, U69135.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ucp2

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s)

response to superoxide
response to hypoxia
transport
mitochondrial transport
positive regulation of cell death
negative regulation of apoptotic process
regulation of mitochondrial membrane potential
negative regulation of insulin secretion involved in cellular response to glucose stimulus

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700012018.mm.1 TC0700001409.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012018.mm.1 NM_011671.5 Refseq
Affymetrix Mouse Clariom S Array PSR0700012022.mm.1 TC0700001409.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012022.mm.1 NM_011671.5 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 535105 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0