This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00627598_m1
See other UCP2 GE Assays ›
Gene Symbol:
Ucp2
Gene Name:
uncoupling protein 2 (mitochondrial, proton carrier)
Gene Aliases:
Slc25a8
Chromosome Location:
Chr.7: 100493358 - 100499630 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00627598_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ucp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011671.5 NP_035801.3 2 - 3 272 89
GenBank mRNA AK035298.1 - 267 89
AK054060.1 - 273 89
AK134116.1 - 2 - 3 273 89
AK142687.1 - 2 - 3 273 89
AK165245.1 - 2 - 3 273 89
BC012697.1 - 2 - 3 271 4205625 89
BC012967.1 - 1 - 2 173 2811421 89
U69135.1 - 2 - 3 200 89
U94593.1 - 2 - 3 193 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK035298.1, AK054060.1, AK134116.1, AK142687.1, BC012697.1, BC012967.1, NM_011671.5, U69135.1, U94593.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ucp2

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s)

response to superoxide
response to hypoxia
transport
mitochondrial transport
positive regulation of cell death
negative regulation of apoptotic process
regulation of mitochondrial membrane potential
negative regulation of insulin secretion involved in cellular response to glucose stimulus

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700012026.mm.1 TC0700001409.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012026.mm.1 NM_011671.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0