This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00627955_g1
See other RNMT GE Assays ›
Gene Symbol:
Rnmt
Gene Name:
RNA (guanine-7-) methyltransferase
Gene Aliases:
2610002P10Rik, AI848273, Rg7mt1, mKIAA0398
Chromosome Location:
Chr.18: 68300338 - 68324852 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00627955_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rnmt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026440.4 NP_080716.1 6 - 7 1052 154
GenBank mRNA AK011300.1 - 6 - 7 1036 154
AK031780.1 - 6 - 7 1010 154
AK082331.1 - 6 - 7 1019 154
AK129130.1 - 5 - 6 989 154
BC021794.1 - 6 - 7 998 4208274 154

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000727.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rnmt

Panther Classification:

Molecular Function -

RNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

7-methylguanosine mRNA capping
mRNA processing
methylation
RNA (guanine-N7)-methylation
mRNA methylation
RNA binding
mRNA (guanine-N7-)-methyltransferase activity
methyltransferase activity
transferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800005314.mm.1 NM_026440.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005314.mm.1 NM_026440.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800005314.mm.1 TC1800000727.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0