This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00656450_m1
See other IAH1 GE Assays ›
Gene Symbol:
Iah1
Gene Name:
isoamyl acetate-hydrolyzing esterase 1 homolog (S. cerevisiae)
Gene Aliases:
1500019E20Rik, 4833421E05Rik
Chromosome Location:
Chr.12: 21316360 - 21323607 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00656450_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Iah1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026347.3 NP_080623.2 5 - 6 612 58
GenBank mRNA AI837665.1 - 1 - 2 58
AK005287.1 - 5 - 6 613 58
BC060949.1 - 5 - 6 615 6533986 58
BC087901.1 - 5 - 6 583 30303437 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000183.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Iah1

Panther Classification:

Molecular Function -

esterase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
biological_process
lipid catabolic process
molecular_function
nuclease activity
hydrolase activity
hydrolase activity, acting on ester bonds
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28280005 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28280008 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28280007 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P507305 AK014743.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P507305 AK005287.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P507305 AK011299.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P507305 BC060949.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P507305 BC087901.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P507305 AK014743.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P507305 AK005287.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P507305 AK011299.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P507305 BC060949.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P507305 BC087901.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26503 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84207 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598226 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295611 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P507305 AK014743.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P507305 AK005287.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P507305 AK011299.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P507305 BC060949.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P507305 BC087901.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P507305 AK014743.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P507305 AK005287.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P507305 AK011299.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P507305 BC060949.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P507305 BC087901.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280016 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279998 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280013 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280015 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280005 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279997 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280020 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280018 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280017 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280023 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280007 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280004 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280002 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280021 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280010 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280003 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280024 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279999 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280009 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28280011 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07556790 - -
Whole Mouse Genome G4122A A_51_P281637 AK014743.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P507305 AK014743.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P507305 AK005287.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P507305 AK011299.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P507305 BC060949.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P507305 BC087901.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P507305 AK014743.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P507305 AK005287.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P507305 AK011299.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P507305 BC060949.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P507305 BC087901.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130353 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109061 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10282980 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10091841 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10282989 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10282975 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10282993 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28280005 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28280008 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28280007 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280043 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280045 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280013 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280024 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279998 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280049 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280011 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280001 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280015 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280005 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280036 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280041 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280040 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280018 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280020 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280009 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280003 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280031 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280021 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280004 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280025 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280046 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280002 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280016 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280010 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280048 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280007 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280017 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280042 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280054 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280034 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280051 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280038 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28280030 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P10282996 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280012 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P10237084 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P10282975 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P10282978 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09823743 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P10282112 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280011 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280040 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P10282990 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280047 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280034 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P10091841 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280038 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280020 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280024 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09962535 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280009 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280048 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09933379 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P10282972 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P10282980 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280031 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280026 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09821516 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280017 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28279997 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280016 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P10282976 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280002 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P10402216 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09821510 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280051 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280021 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28280003 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P10282111 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P10237083 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10394915 - -
Affymetrix Mouse Gene 1.0 ST Array 10394917 - -
Affymetrix Mouse Gene 1.0 ST Array 10394916 - -
Affymetrix Mouse Gene 1.0 ST Array 10394918 - -
Affymetrix Mouse Gene 1.0 ST Array 10394920 - -
Affymetrix Mouse Gene 1.0 ST Array 10394921 - -
Affymetrix Mouse Gene 1.0 ST Array 10394919 - -
Affymetrix HT MG-430 PM Array Plate 1429119_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1453471_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1454898_PM_s_at - -
Affymetrix Mouse Gene 1.1 ST Array 10394914 - -
Mouse Genome 430 2.0 Array 1453471_PM_at - -
Mouse Genome 430 2.0 Array 1429119_at - -
Mouse Genome 430 2.0 Array 1454898_PM_s_at - -
Mouse Genome 430 2.0 Array 1454898_s_at - -
Mouse Genome 430 2.0 Array 1453471_at - -
Mouse Genome 430 2.0 Array 1429119_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67732_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67732_PM_at - -
Affymetrix Mouse Clariom S Array PSR1200001472.mm.1 NM_026347.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200001472.mm.1 NM_026347.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200001472.mm.1 TC1200000183.mm.1 MTA TC
Mouse Expression Set 430 1454898_s_at - -
Mouse Expression Set 430 1454898_PM_s_at - -
Mouse Expression Set 430 1453471_at - -
Mouse Expression Set 430 1453471_PM_at - -
Murine Genome U74v2 Set 167919_PM_at - -
Murine Genome U74v2 Set 167919_at - -
Mouse Genome 430A 2.0 Array 1454898_PM_s_at - -
Mouse Genome 430A 2.0 Array 1454898_s_at - -
Affymetrix HT Mouse Genome 430A Array 1454898_s_at - -
Affymetrix HT Mouse Genome 430A Array 1454898_PM_s_at - -
Affymetrix HT Mouse Genome 430B Array 1429119_at - -
Affymetrix HT Mouse Genome 430B Array 1429119_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453471_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453471_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 915218 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 2900170 - -
Illumina mouse-6 v1.0 expression beadchip 2900170 - -
Illumina mouse-6 v1.0 expression beadchip 1690309 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001690309 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002900170 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2723208 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2620803 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2620803 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2723208 - -
mouse-6 v1.1 (Illumina) scl067732.2_80-S - -
mouse-6 v1.1 (Illumina) scl0067732.1_253-S - -
Illumina Sentrix mouse-6.v1 bead array 1690309 - -
Illumina Sentrix mouse-6.v1 bead array 2900170 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0067732.1_253-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl067732.2_80-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1690309 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2900170 - -
MouseRef-8 v2 Expression BeadChip ILMN_2620803 - -
MouseRef-8 v2 Expression BeadChip ILMN_2620804 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2620804 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2620803 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067732.2_80-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0067732.1_253-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0