This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00658737_m1
See other RGS6 GE Assays ›
Gene Symbol:
Rgs6
Gene Name:
regulator of G-protein signaling 6
Gene Aliases:
-
Chromosome Location:
Chr.12: 82616176 - 83158815 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00658737_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rgs6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282061.1 NP_001268990.1 12 - 13 937 134
NM_015812.3 NP_056627.1 12 - 13 1219 134
GenBank mRNA AF061933.1 - 9 - 10 664 134
AK052527.1 - 12 - 13 1220 134
AK053087.1 - 12 - 13 938 134
AK139323.1 - 12 - 13 1334 134
BC038616.1 - 10 - 11 764 134
BC125029.1 - 12 - 13 923 40062355 134
BC125030.1 - 12 - 13 923 40062356 134

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rgs6

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
negative regulation of signal transduction
intracellular signal transduction
termination of G-protein coupled receptor signaling pathway
positive regulation of GTPase activity
positive regulation of neuron differentiation
signal transducer activity
GTPase activator activity
protein binding
protein domain specific binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0