Assay Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170755.2 | NP_739561.2 | 2 - 3 | 447 | 102 | |
| GenBank mRNA | AK034667.1 | - | 2 - 3 | 448 | 102 | |
| AK152268.1 | - | 2 - 3 | 162 | 102 | ||
| AK158940.1 | - | 2 - 3 | 217 | 102 | ||
| AK160400.1 | - | 2 - 3 | 401 | 102 | ||
| AK165957.1 | - | 2 - 3 | 426 | 102 | ||
| AK172093.1 | - | 2 - 3 | 444 | 102 | ||
| BC070406.1 | - | 2 - 3 | 439 | 5707319 | 102 | |
| BY261213.1 | - | 2 - 3 | 225 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam134a
Genomic Map