This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00661888_m1
See other DGKZ GE Assays ›
Gene Symbol:
Dgkz
Gene Name:
diacylglycerol kinase zeta
Gene Aliases:
E130307B02Rik, ENSMUSG00000075035, F730209L11Rik, mDGK[z]
Chromosome Location:
Chr.2: 91932822 - 91976443 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00661888_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dgkz
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166597.1 NP_001160069.1 26 - 27 3064 109
NM_138306.2 NP_612179.2 25 - 26 2463 109
GenBank mRNA AK087503.1 - 26 - 27 3066 109
AK170936.1 - 22 - 23 2086 109
BC014860.1 - 10 - 11 933 109
BC049228.1 - 25 - 26 2499 3668704 109
BE862117.1 - 2 - 3 192 109
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK088841.1 - Chr. - - [-] on Build 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004130.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dgkz

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
mitotic G1 DNA damage checkpoint
intracellular signal transduction
negative regulation of catalytic activity
negative regulation of mitotic cell cycle
negative regulation of Ras protein signal transduction
lipid phosphorylation
nucleotide binding
lipid kinase activity
diacylglycerol kinase activity
enzyme inhibitor activity
protein binding
ATP binding
protein C-terminus binding
kinase activity
transferase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200034614.mm.1 NM_001166597.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034614.mm.1 NM_001166597.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034614.mm.1 NM_138306.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034614.mm.1 NM_138306.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200034614.mm.1 TC0200004130.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline