This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00661890_g1
See other DGKZ GE Assays ›
Gene Symbol:
Dgkz
Gene Name:
diacylglycerol kinase zeta
Gene Aliases:
E130307B02Rik, ENSMUSG00000075035, F730209L11Rik, mDGK[z]
Chromosome Location:
Chr.2: 91932822 - 91976443 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00661890_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dgkz
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166597.1 NP_001160069.1 28 - 29 3135 82
NM_138306.2 NP_612179.2 27 - 28 2534 82
GenBank mRNA AK087503.1 - 28 - 29 3137 82
AK170936.1 - 24 - 25 2157 82
BC014860.1 - 12 - 13 1004 82
BC049228.1 - 27 - 28 2570 3668704 82
BE862117.1 - 4 - 5 263 82
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK088841.1 - Chr. - - [-] on Build 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004130.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dgkz

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
mitotic G1 DNA damage checkpoint
intracellular signal transduction
negative regulation of catalytic activity
negative regulation of mitotic cell cycle
negative regulation of Ras protein signal transduction
lipid phosphorylation
nucleotide binding
lipid kinase activity
diacylglycerol kinase activity
enzyme inhibitor activity
protein binding
ATP binding
protein C-terminus binding
kinase activity
transferase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200034610.mm.1 NM_001166597.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034610.mm.1 NM_001166597.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034610.mm.1 NM_138306.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034610.mm.1 NM_138306.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200034610.mm.1 TC0200004130.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline