This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00675633_g1
See other PDK2 GE Assays ›
Gene Symbol:
Pdk2
Gene Name:
pyruvate dehydrogenase kinase, isoenzyme 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 95026258 - 95041371 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00675633_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdk2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133667.2 NP_598428.2 8 - 9 979 78
GenBank mRNA AK002647.1 - 8 - 9 923 78
BC021764.1 - 8 - 9 933 3488813 78
CK618575.1 - 1 - 2 34 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003583.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pdk2

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glucose metabolic process
regulation of gluconeogenesis
protein phosphorylation
regulation of pH
insulin receptor signaling pathway
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of cellular ketone metabolic process
regulation of glucose metabolic process
phosphorylation
peptidyl-serine phosphorylation
cellular response to nutrient
cellular response to reactive oxygen species
glucose homeostasis
intrinsic apoptotic signaling pathway by p53 class mediator
nucleotide binding
protein serine/threonine kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
protein binding
ATP binding
kinase activity
transferase activity
protein complex binding
protein homodimerization activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100032836.mm.1 NM_133667.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032836.mm.1 NM_133667.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100032836.mm.1 TC1100003583.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100032838.mm.1 NM_133667.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032838.mm.1 NM_133667.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100032838.mm.1 TC1100003583.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0