| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700008660.mm.1 |
NM_008071.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008660.mm.1 |
NM_008071.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008660.mm.1 |
TC0700000941.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700008663.mm.1 |
NM_001038701.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008663.mm.1 |
NM_008071.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008663.mm.1 |
NM_008071.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008663.mm.1 |
TC0700000941.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700008662.mm.1 |
NM_001038701.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008662.mm.1 |
NM_008071.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008662.mm.1 |
NM_008071.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008662.mm.1 |
TC0700000941.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700008661.mm.1 |
NM_008071.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008661.mm.1 |
NM_008071.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008661.mm.1 |
TC0700000941.mm.1 |
MTA TC |