Assay Details
| Gene Symbol: Trps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC037058.1 | - | 5 - 6 | 2179 | 4951294 | 66 |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trps1| Gene Symbol: Trps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC037058.1 | - | 5 - 6 | 2179 | 4951294 | 66 |
Set Membership: |
|