This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00712572_m1
See other MS4A7 GE Assays ›
Gene Symbol:
Ms4a7
Gene Name:
membrane-spanning 4-domains, subfamily A, member 7
Gene Aliases:
9130422I10Rik, A430103C15Rik, CD20l4, CFFMA
Chromosome Location:
Chr.19: 11321039 - 11336146 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00712572_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ms4a7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025610.4 NP_001020781.1 4 - 5 631 149
NM_001276398.1 NP_001263327.1 5 - 6 928 149
NM_027836.6 NP_082112.3 5 - 6 688 149
GenBank mRNA AF237917.2 - 5 - 6 639 149
AK018686.1 - 4 - 5 602 149
AK079888.1 - 5 - 6 929 149
AK149890.1 - 5 - 6 656 149
AK151261.1 - 5 - 6 659 149
AK152098.1 - 5 - 6 659 149

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001131.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ms4a7

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900010639.mm.1 NM_001025610.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900010639.mm.1 NM_001276398.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900010639.mm.1 NM_027836.6 Refseq
Affymetrix Mouse Clariom S Array PSR1900010639.mm.1 TC1900001131.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0