This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00712778_m1
See other DTL GE Assays ›
Gene Symbol:
Dtl
Gene Name:
denticleless homolog (Drosophila)
Gene Aliases:
2810047L02Rik, 5730564G15Rik, Ramp
Chromosome Location:
Chr.1: 191537365 - 191575534 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00712778_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dtl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK054412.1 - 12 - 13 1234 71
AK133177.1 - 12 - 13 1243 71
AK154594.1 - 13 - 14 1284 71
AK161401.1 - 9 - 10 845 71
AK164249.1 - 12 - 13 1244 71
AK166351.1 - 12 - 13 1252 71
AK169601.1 - 12 - 13 1243 71
BC060208.1 - 12 - 13 1203 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline