This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00712788_m1
See other DTL GE Assays ›
Gene Symbol:
Dtl
Gene Name:
denticleless homolog (Drosophila)
Gene Aliases:
2810047L02Rik, 5730564G15Rik, Ramp
Chromosome Location:
Chr.1: 191537365 - 191575534 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00712788_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dtl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB095735.1 - 8 - 9 828 99
AB095736.1 - 8 - 9 828 99
AK054412.1 - 8 - 9 818 99
AK133177.1 - 8 - 9 827 99
AK154594.1 - 9 - 10 868 99
AK161401.1 - 5 - 6 429 99
AK164249.1 - 8 - 9 828 99
AK164999.1 - 8 - 9 827 99
AK166351.1 - 8 - 9 836 99
AK169601.1 - 8 - 9 827 99
BC060208.1 - 8 - 9 787 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline