This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00712987_g1
See other IGSF8 GE Assays ›
Gene Symbol:
Igsf8
Gene Name:
immunoglobulin superfamily, member 8
Gene Aliases:
AA033172, AU041109, ESTM34, EWI-2, KCT-4, PGRL, R75384
Chromosome Location:
Chr.1: 172312407 - 172319841 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00712987_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080419.1 NP_536344.1 5 - 6 1853 128
GenBank mRNA AF411055.1 - 5 - 6 1724 128
AF439263.1 - 5 - 6 1836 128
AK140254.1 - 5 - 6 1865 128
AK146104.1 - 5 - 6 1866 128
AK146221.1 - 5 - 6 1872 128
AK158522.1 - 5 - 6 1874 128
AK168363.1 - 5 - 6 1872 128
BC019587.1 - 3 - 4 875 128
BC026464.1 - 4 - 5 1659 128
BC026633.1 - 1 - 2 405 128
BC038068.1 - 3 - 4 1110 128
BC048387.1 - 5 - 6 1853 6311441 128
BC060087.1 - 2481 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001610.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igsf8

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell motility
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100013390.mm.1 NM_080419.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100013390.mm.1 TC0100001610.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100013389.mm.1 NM_080419.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100013389.mm.1 TC0100001610.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100013388.mm.1 NM_080419.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100013388.mm.1 TC0100001610.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline