This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00723305_m1
See other HMGCLL1 GE Assays ›
Gene Symbol:
Hmgcll1
Gene Name:
3-hydroxymethyl-3-methylglutaryl-Coenzyme A lyase-like 1
Gene Aliases:
BC037381
Chromosome Location:
Chr.9: 76014650 - 76136350 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00723305_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hmgcll1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173731.2 NP_776092.1 7 - 8 890 78
GenBank mRNA AK141622.1 - 7 - 8 853 78
BC037381.1 - 7 - 8 890 5368383 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001022.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hmgcll1

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

ketone body biosynthetic process
catalytic activity
hydroxymethylglutaryl-CoA lyase activity
lyase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900008089.mm.1 NM_173731.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900008089.mm.1 NM_173731.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900008089.mm.1 TC0900001022.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0