This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00724383_g1
See other KAT5 GE Assays ›
Gene Symbol:
Kat5
Gene Name:
K(lysine) acetyltransferase 5
Gene Aliases:
AI839539, CPLA2, Htatip, Htatip1, PLIP, Tip55, Tip60
Chromosome Location:
Chr.19: 5603014 - 5610232 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00724383_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kat5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199247.1 NP_001186176.1 7 - 8 754 58
NM_001199248.1 NP_001186177.1 6 - 7 853 58
NM_001199249.1 NP_001186178.1 6 - 7 853 58
NM_178637.2 NP_848752.1 8 - 9 910 58
NR_037603.1 5 - 6 786 58
GenBank mRNA AB055409.1 - 5 - 6 764 58
AF528194.1 - 4 - 5 464 58
AF528195.1 - 7 - 8 845 58
AF528196.1 - 6 - 7 689 58
AK143674.1 - 6 - 7 805 58
AK146352.1 - 8 - 9 867 58
AK159568.1 - 6 - 7 786 58
AK160077.1 - 6 - 7 786 58
AK219598.1 - 1 - 2 131 58
BC110675.1 - 8 - 9 851 4011689 58
BC129967.1 - 6 - 7 688 58
BC129968.1 - 6 - 7 689 40106268 58
CA317697.1 - 1 - 2 148 58
DQ139980.1 - 7 - 8 845 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000975.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Kat5

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
response to ionizing radiation
chromatin modification
histone acetylation
negative regulation of interleukin-2 production
regulation of growth
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cellular response to estradiol stimulus
positive regulation of protein acetylation
chromatin binding
transcription coactivator activity
histone acetyltransferase activity
protein binding
transcription factor binding
transferase activity
transferase activity, transferring acyl groups
transferase activity, transferring acyl groups other than amino-acyl groups
protein complex binding
phospholipase binding
metal ion binding
repressing transcription factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900008700.mm.1 NM_001199247.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900008700.mm.1 NM_001199247.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900008700.mm.1 NM_001199248.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900008700.mm.1 NM_001199248.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900008700.mm.1 NM_001199249.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900008700.mm.1 NM_001199249.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900008700.mm.1 NM_178637.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900008700.mm.1 NM_178637.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900008700.mm.1 NR_037603.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900008700.mm.1 NR_037603.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900008700.mm.1 TC1900000975.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0