This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00725381_m1
See other SDR16C5 GE Assays ›
Gene Symbol:
Sdr16c5
Gene Name:
short chain dehydrogenase/reductase family 16C, member 5
Gene Aliases:
Rdhe2, Scdr9
Chromosome Location:
Chr.4: 3995942 - 4019663 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00725381_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sdr16c5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181989.1 NP_871789.1 5 - 6 935 131
GenBank mRNA AK136508.1 - 4 - 5 906 131
AY277588.1 - 5 - 6 935 131
BC104134.1 - 5 - 6 854 40045268 131
BC104135.1 - 5 - 6 854 40045269 131

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002136.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sdr16c5

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
retinol metabolic process
retinal metabolic process
keratinocyte proliferation
oxidation-reduction process
retinol dehydrogenase activity
oxidoreductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400017963.mm.1 NM_181989.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017963.mm.1 NM_181989.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017963.mm.1 TC0400002136.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0