This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00725708_g1
See other GLUL GE Assays ›
Gene Symbol:
Glul
Gene Name:
glutamate-ammonia ligase (glutamine synthetase)
Gene Aliases:
GS, Glns
Chromosome Location:
Chr.1: 153899929 - 153909723 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00725708_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glul
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008131.4 NP_032157.2 5 669 61
GenBank mRNA AK137969.1 - 5 656 61
AK159106.1 - 5 655 61
AK160670.1 - 5 656 61
AK162685.1 - 5 654 61
AK168493.1 - 5 653 61
AY044241.1 - 5 653 61
BC015086.1 - 5 653 4224865 61
U09114.1 - 4 545 61
X16314.1 - 5 630 61

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Glul

Panther Classification:

Molecular Function -

ligase

Gene Ontology Categories:

Function(s) Process(es)

glutamate metabolic process
glutamine biosynthetic process
nitrogen compound metabolic process
cell proliferation
cellular response to starvation
response to glucose
ammonia assimilation cycle
positive regulation of insulin secretion
positive regulation of epithelial cell proliferation
protein homooligomerization
positive regulation of synaptic transmission, glutamatergic
nucleotide binding
magnesium ion binding
catalytic activity
glutamate decarboxylase activity
glutamate-ammonia ligase activity
protein binding
ATP binding
glutamate binding
lyase activity
ligase activity
manganese ion binding
identical protein binding
dynein light chain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0