This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00725978_g1
See other TWF1 GE Assays ›
Gene Symbol:
Twf1
Gene Name:
twinfilin, actin-binding protein, homolog 1 (Drosophila)
Gene Aliases:
A6, Ptk9, twinfilin
Chromosome Location:
Chr.15: 94577948 - 94589884 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00725978_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Twf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008971.4 NP_032997.3 2 - 3 149 75
GenBank mRNA AK077410.1 - 2 - 3 205 75
AK147990.1 - 2 - 3 143 75
AK150852.1 - 2 - 3 172 75
AK168142.1 - 2 - 3 167 75
AK168161.1 - 2 - 3 167 75
AY267188.1 - 2 - 3 101 75
BC015081.1 - 2 - 3 166 3979221 75
BC094034.1 - 2 - 3 149 4934593 75
U82324.1 - 2 - 3 157 75

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Twf1

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-tyrosine phosphorylation
negative regulation of actin filament polymerization
sequestering of actin monomers
regulation of actin phosphorylation
barbed-end actin filament capping
actin binding
actin monomer binding
protein tyrosine kinase activity
ATP binding
phosphatidylinositol-4,5-bisphosphate binding
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500016990.mm.1 NM_008971.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016990.mm.1 NM_008971.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500016990.mm.1 TC1500002111.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0