This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00727680_s1
See other GCNT2 GE Assays ›
Gene Symbol:
Gcnt2
Gene Name:
glucosaminyl (N-acetyl) transferase 2, I-branching enzyme
Gene Aliases:
5330430K10Rik, IGnT, IGnTA, IGnTB, IGnTC
Chromosome Location:
Chr.13: 40859768 - 40960892 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00727680_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gcnt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023887.4 NP_076376.3 1 1346 74
GenBank mRNA AK156328.1 - 1 882 74
AY236874.1 - 1 1146 74
AY435148.1 - 1 759 74
BC094572.1 - 1 1346 30356538 74
BQ892107.1 - 1 517 74

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gcnt2

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
transforming growth factor beta receptor signaling pathway
positive regulation of cell proliferation
posttranscriptional regulation of gene expression
positive regulation of epithelial to mesenchymal transition
negative regulation of cell-substrate adhesion
positive regulation of cell migration
positive regulation of heterotypic cell-cell adhesion
maintenance of lens transparency
positive regulation of protein kinase B signaling
positive regulation of ERK1 and ERK2 cascade
N-acetyllactosaminide beta-1,6-N-acetylglucosaminyltransferase activity
acetylglucosaminyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300002713.mm.1 NM_023887.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300002713.mm.1 NM_023887.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300002713.mm.1 TC1300000460.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 428470 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0