This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00769589_m1
See other FRS2 GE Assays ›
Gene Symbol:
Frs2
Gene Name:
fibroblast growth factor receptor substrate 2
Gene Aliases:
4732458E18, C330018A15Rik, Frs2alpha, SNT1
Chromosome Location:
Chr.10: 117070127 - 117148474 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00769589_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Frs2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177798.3 NP_808466.1 4 - 5 230 146
GenBank mRNA AK028813.1 - 4 - 5 246 146
AK163442.1 - 4 - 5 252 146
BC043109.1 - 4 - 5 230 6406912 146
BC055334.1 - 4 - 5 266 6830555 146

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK028813.1, BC043109.1, BC055334.1, NM_177798.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Frs2

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
gastrulation with mouth forming second
organ induction
lens development in camera-type eye
ventricular septum development
transmembrane receptor protein tyrosine kinase signaling pathway
neuroblast proliferation
fibroblast growth factor receptor signaling pathway
anterior/posterior axis specification, embryo
positive regulation of signal transduction
forebrain development
regulation of apoptotic process
optic placode formation involved in camera-type eye formation
regulation of epithelial cell proliferation
prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
lens fiber cell development
regulation of ERK1 and ERK2 cascade
transmembrane receptor protein tyrosine kinase adaptor activity
fibroblast growth factor receptor binding
insulin receptor binding
neurotrophin TRKA receptor binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline