This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00770942_m1
See other POLR2D GE Assays ›
Gene Symbol:
Polr2d
Gene Name:
polymerase (RNA) II (DNA directed) polypeptide D
Gene Aliases:
2310002G05Rik, 2610028L19Rik, HSRBP4, RBP4
Chromosome Location:
Chr.18: 31789159 - 31796701 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00770942_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr2d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027002.3 NP_081278.1 1 - 2 85 83
NM_027101.2 NP_081377.1 1 - 2 68 83
GenBank mRNA AK009093.1 - 1 - 2 89 83
AK011600.1 - 1 - 2 87 83

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_027101.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polr2d

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
mRNA export from nucleus in response to heat stress
recruitment of 3'-end processing factors to RNA polymerase II holoenzyme complex
cellular metabolic process
positive regulation of translational initiation
nucleotide binding
single-stranded DNA binding
single-stranded RNA binding
catalytic activity
DNA-directed RNA polymerase activity
translation initiation factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z8r28:80/100.66.79.24:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0