This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00772675_g1
See other RBPMS2 GE Assays ›
Gene Symbol:
Rbpms2
Gene Name:
RNA binding protein with multiple splicing 2
Gene Aliases:
2400008B06Rik, AI316523
Chromosome Location:
Chr.9: 65629336 - 65660536 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00772675_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbpms2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028030.3 NP_082306.2 6 - 7 807 111
GenBank mRNA AK144659.1 - 6 - 7 832 111
AK156633.1 - 6 - 7 807 111
BC021788.1 - 6 - 7 805 4236661 111

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbpms2

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
molecular_function
nucleic acid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900006505.mm.1 NM_028030.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006505.mm.1 NM_028030.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006505.mm.1 TC0900000848.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006506.mm.1 NM_028030.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006506.mm.1 NM_028030.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006506.mm.1 TC0900000848.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0