This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00802030_m1
See other ABL1 GE Assays ›
Gene Symbol:
Abl1
Gene Name:
c-abl oncogene 1, non-receptor tyrosine kinase
Gene Aliases:
AI325092, Abl, E430008G22Rik, c-Abl
Chromosome Location:
Chr.2: 31688252 - 31807093 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00802030_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009594.4 NP_033724.2 1 - 2 180 89
GenBank mRNA AK090095.1 - 1 - 2 138 89
CJ099623.1 - 1 - 2 131 89
CJ128107.1 - 1 - 2 175 89
J02995.1 - 1 - 2 133 89
K03228.1 - 1 - 2 175 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000558.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Abl1

Gene Ontology Categories:

Function(s) Process(es)

B-1 B cell homeostasis
positive regulation of protein phosphorylation
B cell proliferation involved in immune response
transitional one stage B cell differentiation
DNA repair
protein phosphorylation
endocytosis
phagocytosis
autophagy
apoptotic process
cellular response to DNA damage stimulus
DNA damage induced protein phosphorylation
response to oxidative stress
cell adhesion
epidermal growth factor receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
phosphorylation
cell migration
peptidyl-tyrosine phosphorylation
cerebellum morphogenesis
negative regulation of cell-cell adhesion
microspike assembly
actin cytoskeleton organization
negative regulation of BMP signaling pathway
regulation of actin cytoskeleton organization
positive regulation of osteoblast proliferation
substrate adhesion-dependent cell spreading
peptidyl-tyrosine autophosphorylation
B cell proliferation
signal transduction in response to DNA damage
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
negative regulation of mitotic cell cycle
positive regulation of mitotic cell cycle
alpha-beta T cell differentiation
platelet-derived growth factor receptor signaling pathway
spleen development
thymus development
collateral sprouting
positive regulation of peptidyl-tyrosine phosphorylation
activated T cell proliferation
B cell receptor signaling pathway
neuromuscular process controlling balance
positive regulation of release of sequestered calcium ion into cytosol
positive regulation of oxidoreductase activity
negative regulation of ubiquitin-protein transferase activity
regulation of cell cycle
Bergmann glial cell differentiation
negative regulation of ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
cellular response to lipopolysaccharide
negative regulation of protein serine/threonine kinase activity
actin filament branching
positive regulation of interleukin-2 secretion
negative regulation of phospholipase C activity
positive regulation of neuron death
positive regulation of interferon-gamma secretion
regulation of extracellular matrix organization
positive regulation of Wnt signaling pathway, planar cell polarity pathway
negative regulation of endothelial cell apoptotic process
regulation of cellular senescence
negative regulation of cellular senescence
regulation of response to DNA damage stimulus
nucleotide binding
magnesium ion binding
DNA binding
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
protein C-terminus binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
SH3 domain binding
protein kinase binding
protein domain specific binding
syntaxin binding
manganese ion binding
metal ion binding
actin filament binding
mitogen-activated protein kinase binding
proline-rich region binding
delta-catenin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200004826.mm.1 NM_009594.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200004826.mm.1 TC0200000558.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200004829.mm.1 NM_001112703.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200004829.mm.1 NM_001283045.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200004829.mm.1 NM_001283046.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200004829.mm.1 NM_001283047.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200004829.mm.1 NM_009594.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200004829.mm.1 TC0200000558.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200004830.mm.1 NM_001112703.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200004830.mm.1 NM_001283045.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200004830.mm.1 NM_001283046.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200004830.mm.1 NM_001283047.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200004830.mm.1 NM_009594.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200004830.mm.1 TC0200000558.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0