This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00802040_m1
See other ACE GE Assays ›
Gene Symbol:
Ace
Gene Name:
angiotensin I converting enzyme (peptidyl-dipeptidase A) 1
Gene Aliases:
AW208573, CD143
Chromosome Location:
Chr.11: 105967945 - 105989964 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00802040_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ace
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001281819.1 NP_001268748.1 22 - 23 3427 74
NM_009598.2 NP_033728.1 11 - 12 1650 74
NM_207624.5 NP_997507.1 22 - 23 3427 74
GenBank mRNA AK154632.1 - 22 - 23 3426 74
AK161020.1 - 22 - 23 3423 74
AK164780.1 - 9 - 10 1295 74
BC034367.1 - 18 - 19 2689 74
BC040404.1 - 22 - 23 3416 3965649 74
BC052531.1 - 3 - 4 279 74
BC083109.1 - 22 - 23 3205 74
BC110362.1 - 22 - 23 3413 5343978 74
J04946.1 - 16 - 17 2394 74
J04947.1 - 2394 74
M55333.1 - 11 - 12 1650 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ace

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

kidney development
neutrophil mediated immunity
regulation of systemic arterial blood pressure by renin-angiotensin
positive regulation of systemic arterial blood pressure
proteolysis
peptide metabolic process
spermatogenesis
regulation of blood pressure
posttranscriptional regulation of gene expression
negative regulation of gene expression
bradykinin catabolic process
regulation of smooth muscle cell migration
sensory perception of pain
organ regeneration
negative regulation of protein binding
positive regulation of protein binding
response to lipopolysaccharide
vasoconstriction
hormone catabolic process
eating behavior
positive regulation of apoptotic process
peptide catabolic process
lung alveolus development
beta-amyloid metabolic process
arachidonic acid secretion
positive regulation of inflammatory response
positive regulation of neurogenesis
heart contraction
regulation of angiotensin metabolic process
angiogenesis involved in coronary vascular morphogenesis
positive regulation of protein tyrosine kinase activity
cell proliferation in bone marrow
negative regulation of calcium ion import
positive regulation of peptidyl-tyrosine autophosphorylation
regulation of hematopoietic stem cell proliferation
negative regulation of gap junction assembly
positive regulation of peptidyl-cysteine S-nitrosylation
actin binding
endopeptidase activity
carboxypeptidase activity
protein binding
drug binding
peptidase activity
metallopeptidase activity
exopeptidase activity
tripeptidyl-peptidase activity
peptidyl-dipeptidase activity
zinc ion binding
lytic transglycosylase activity
mannosidase activity
mannosyl-oligosaccharide mannosidase activity
galactosidase activity
glucosidase activity
trehalase activity
fucosidase activity
hexosaminidase activity
amylase activity
hydrolase activity
chloride ion binding
mitogen-activated protein kinase kinase binding
bradykinin receptor binding
dextrin alpha-glucosidase activity
starch alpha-glucosidase activity
metal ion binding
mitogen-activated protein kinase binding
beta-glucanase activity
beta-6-sulfate-N-acetylglucosaminidase activity
glucan endo-1,4-beta-glucosidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline