This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00802106_g1
See other APBB1 GE Assays ›
Gene Symbol:
Apbb1
Gene Name:
amyloid beta (A4) precursor protein-binding, family B, member 1
Gene Aliases:
Fe65, Rir
Chromosome Location:
Chr.7: 105558465 - 105581653 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00802106_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apbb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253885.1 NP_001240814.1 13 - 14 2159 82
NM_001253886.1 NP_001240815.1 12 - 13 2153 82
NM_001253887.1 NP_001240816.1 13 - 14 1159 82
NM_009685.3 NP_033815.1 12 - 13 1967 82
GenBank mRNA AK030748.1 - 12 - 13 1850 82
AK075730.1 - 13 - 14 1139 82
AK083830.1 - 13 - 14 1889 82
AK144340.1 - 13 - 14 2160 82
AK164140.1 - 13 - 14 1884 82
BC022732.1 - 6 - 7 534 82
BC036956.1 - 12 - 13 1797 82
BC048395.1 - 11 - 12 1305 6392016 82
L77865.1 - 3 - 4 291 82

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apbb1

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
cellular response to DNA damage stimulus
cell cycle arrest
axon guidance
visual learning
chromatin modification
actin filament-based movement
extracellular matrix organization
negative regulation of cell growth
positive regulation of apoptotic process
histone H4 acetylation
negative regulation of neuron differentiation
positive regulation of DNA repair
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein secretion
negative regulation of thymidylate synthase biosynthetic process
protein stabilization
beta-amyloid binding
chromatin binding
protein binding
transcription factor binding
protein complex binding
histone acetyltransferase binding
histone binding
proline-rich region binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0