This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00802112_g1
See other APBB1 GE Assays ›
Gene Symbol:
Apbb1
Gene Name:
amyloid beta (A4) precursor protein-binding, family B, member 1
Gene Aliases:
Fe65, Rir
Chromosome Location:
Chr.7: 105558465 - 105581653 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00802112_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apbb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253885.1 NP_001240814.1 7 - 8 1629 112
NM_001253886.1 NP_001240815.1 7 - 8 1629 112
NM_001253887.1 NP_001240816.1 7 - 8 629 112
NM_009685.3 NP_033815.1 7 - 8 1443 112
GenBank mRNA AK030748.1 - 7 - 8 1326 112
AK075730.1 - 7 - 8 609 112
AK083830.1 - 7 - 8 1359 112
AK144340.1 - 7 - 8 1630 112
AK164140.1 - 7 - 8 1354 112
BC036956.1 - 6 - 7 1267 112
BC048395.1 - 5 - 6 775 6392016 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003981.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apbb1

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
cellular response to DNA damage stimulus
cell cycle arrest
axon guidance
visual learning
chromatin modification
actin filament-based movement
extracellular matrix organization
negative regulation of cell growth
positive regulation of apoptotic process
histone H4 acetylation
negative regulation of neuron differentiation
positive regulation of DNA repair
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein secretion
negative regulation of thymidylate synthase biosynthetic process
protein stabilization
beta-amyloid binding
chromatin binding
protein binding
transcription factor binding
protein complex binding
histone acetyltransferase binding
histone binding
proline-rich region binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700033127.mm.1 NM_001253885.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033127.mm.1 NM_001253885.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033127.mm.1 NM_001253886.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033127.mm.1 NM_001253886.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033127.mm.1 NM_001253887.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033127.mm.1 NM_001253887.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033127.mm.1 NM_009685.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033127.mm.1 NM_009685.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700033127.mm.1 TC0700003981.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jrd88:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0