This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00802124_m1
See other APBB2 GE Assays ›
Gene Symbol:
Apbb2
Gene Name:
amyloid beta (A4) precursor protein-binding, family B, member 2
Gene Aliases:
2310007D03Rik, FE65L1, Rirl1, TR2L, Zfra
Chromosome Location:
Chr.5: 66298725 - 66618817 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00802124_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apbb2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001201413.1 NP_001188342.1 5 - 6 1403 85
NM_001201415.1 NP_001188344.1 5 - 6 1403 85
NM_009686.2 NP_033816.1 5 - 6 1403 85
GenBank mRNA AK004792.1 - 5 - 6 1257 85
AK167649.1 - 5 - 6 1268 85
AK169495.1 - 5 - 6 1256 85
AK169959.1 - 5 - 6 1344 85
BC076587.1 - 5 - 6 1249 30550087 85

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apbb2

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
regulation of transcription, DNA-templated
cell cycle arrest
axon guidance
actin filament-based movement
extracellular matrix organization
negative regulation of cell growth
positive regulation of apoptotic process
negative regulation of apoptotic process
protein stabilization
beta-amyloid binding
protein binding
transcription factor binding
histone acetyltransferase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0