This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00802192_m1
See other BCAT2 GE Assays ›
Gene Symbol:
Bcat2
Gene Name:
branched chain aminotransferase 2, mitochondrial
Gene Aliases:
Bcat-2, Eca40
Chromosome Location:
Chr.7: 45569855 - 45589710 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00802192_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bcat2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243052.1 NP_001229981.1 10 - 11 1392 71
NM_001243053.1 NP_001229982.1 11 - 12 1277 71
NM_009737.3 NP_033867.1 10 - 11 1365 71
GenBank mRNA AF031467.1 - 10 - 11 1157 71
AK008981.1 - 1 - 2 68 71
AK137049.1 - 11 - 12 1328 71
AK145300.1 - 10 - 11 1210 71
AK160159.1 - 10 - 11 1393 71
BC017688.1 - 10 - 11 1142 4242224 71
BC048072.1 - 10 - 11 1156 4166264 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000853.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bcat2

Panther Classification:

Molecular Function -

transaminase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

isoleucine metabolic process
isoleucine catabolic process
leucine metabolic process
valine metabolic process
metabolic process
cellular amino acid biosynthetic process
branched-chain amino acid metabolic process
branched-chain amino acid biosynthetic process
branched-chain amino acid catabolic process
leucine biosynthetic process
regulation of hormone levels
catalytic activity
branched-chain-amino-acid transaminase activity
transaminase activity
transferase activity
L-leucine transaminase activity
L-valine transaminase activity
L-isoleucine transaminase activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093841 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093840 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093839 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093838 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093837 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093842 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960167 BC017688.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960167 BC048072.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960167 AK160159.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960167 AK137049.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960167 AK008981.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 888135 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 103907 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455392 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 42966 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240941 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956475 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333993 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154003 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913492 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767953 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 94265 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137577 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33206 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127934 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118076 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960167 BC017688.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960167 BC048072.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960167 AK160159.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960167 AK137049.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960167 AK008981.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P181448 BC017688.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P181448 BC048072.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P181448 AK160159.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P181448 AK137049.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P181448 AK008981.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P448266 BC017688.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P448266 BC048072.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P448266 AK160159.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P448266 AK137049.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P448266 AK145300.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P393781 BC017688.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P393781 BC048072.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P393781 AK160159.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P393781 U68526.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P393781 AK037482.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093851 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093824 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093827 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093832 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093830 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093850 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093828 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093843 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093845 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093847 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093848 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093852 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093839 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093829 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093834 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093825 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093838 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093849 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093837 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093846 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093833 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093854 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093841 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127934 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118076 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01507989 - -
Whole Mouse Genome G4122A A_52_P448266 BC017688.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P448266 BC048072.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P448266 AK160159.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P448266 AK137049.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P448266 AK145300.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P393781 BC017688.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P393781 BC048072.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P393781 AK160159.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P393781 U68526.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P393781 AK037482.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P181448 BC017688.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P181448 BC048072.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P181448 AK160159.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P181448 AK137049.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P181448 AK008981.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103033 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118066 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104692 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118076 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127934 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06535990 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06535991 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093840 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093837 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093838 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093842 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093839 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093841 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093824 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093830 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093962 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093866 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093959 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093813 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093954 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093870 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093916 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093799 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093837 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093796 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093850 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093884 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093944 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093915 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093920 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093935 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093832 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093846 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093807 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093802 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093844 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093851 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093834 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093852 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093868 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093930 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093823 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093838 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093961 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093828 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093864 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093808 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093875 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093940 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093843 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093803 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093862 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093845 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093883 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093810 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093914 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093877 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093848 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093833 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093841 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093825 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093932 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093927 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093922 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093859 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093923 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093956 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093872 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093812 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093950 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093958 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093792 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093804 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093937 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093806 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093847 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093825 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093874 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093834 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093830 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093864 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093863 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093862 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093837 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093850 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093851 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093843 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093844 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06664138 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093839 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093870 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06421100 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093827 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093845 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093854 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06664127 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093828 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06104560 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093824 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06421105 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093866 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06664130 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093838 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06104561 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093829 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093833 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093868 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06326945 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093872 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06664132 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06421104 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093823 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093832 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093857 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093875 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25093877 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10553016 - -
Affymetrix Mouse Gene 1.0 ST Array 10553020 - -
Affymetrix Mouse Gene 1.0 ST Array 10553024 - -
Affymetrix Mouse Gene 1.0 ST Array 10553022 - -
Affymetrix Mouse Gene 1.0 ST Array 10553018 - -
Affymetrix Mouse Gene 1.0 ST Array 10553023 - -
Affymetrix Mouse Gene 1.0 ST Array 10553019 - -
Affymetrix Mouse Gene 1.0 ST Array 10553025 - -
Affymetrix Mouse Gene 1.0 ST Array 10553017 - -
Affymetrix Mouse Gene 1.0 ST Array 10553021 - -
Affymetrix Mouse Gene 1.0 ST Array 10553026 - -
Affymetrix HT MG-430 PM Array Plate 1425764_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10553015 - -
Mouse Genome 430 2.0 Array 1425764_a_at - -
Mouse Genome 430 2.0 Array 1425764_PM_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 12036_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 12036_at - -
Affymetrix Mouse Clariom S Array PSR0700007448.mm.1 NM_001243052.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007448.mm.1 NM_001243052.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007448.mm.1 NM_001243053.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007448.mm.1 NM_001243053.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007448.mm.1 NM_009737.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007448.mm.1 NM_009737.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700007448.mm.1 TC0700000853.mm.1 MTA TC
Mouse Expression Set 430 1425764_a_at - -
Mouse Expression Set 430 1425764_PM_a_at - -
Murine Genome U74v2 Set 100443_PM_at - -
Murine Genome U74v2 Set 100443_at - -
Mouse Genome 430A 2.0 Array 1425764_PM_a_at - -
Mouse Genome 430A 2.0 Array 1425764_a_at - -
Affymetrix HT Mouse Genome 430A Array 1425764_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1425764_a_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102810091 - -
Illumina mouse-6 v1.0 expression beadchip 5890112 - -
Illumina mouse-6 v1.0 expression beadchip 102810091 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005890112 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2607961 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2607961 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2564659 - -
mouse-6 v1.1 (Illumina) scl32674.12.1_0-S - -
Illumina Sentrix mouse-6.v1 bead array 102810091 - -
Illumina Sentrix mouse-6.v1 bead array 5890112 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl32674.12.1_0-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5890112 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 102810091 - -
MouseRef-8 v2 Expression BeadChip ILMN_2794608 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2794608 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2564659 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline