This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00802194_g1
See other BCAT2 GE Assays ›
Gene Symbol:
Bcat2
Gene Name:
branched chain aminotransferase 2, mitochondrial
Gene Aliases:
Bcat-2, Eca40
Chromosome Location:
Chr.7: 45569855 - 45589710 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00802194_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bcat2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243052.1 NP_001229981.1 2 - 3 351 78
NM_001243053.1 NP_001229982.1 3 - 4 236 78
NM_009737.3 NP_033867.1 2 - 3 324 78
GenBank mRNA AF031467.1 - 2 - 3 116 78
AK037482.1 - 2 - 3 129 78
AK137049.1 - 3 - 4 287 78
AK145300.1 - 2 - 3 169 78
AK160159.1 - 2 - 3 352 78
BB630667.1 - 2 - 3 127 78
BC017688.1 - 2 - 3 101 4242224 78
BC048072.1 - 2 - 3 115 4166264 78
U68526.1 - 2 - 3 110 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF031467.1, AK137049.1, AK160159.1, NM_001243052.1, NM_001243053.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bcat2

Panther Classification:

Molecular Function -

transaminase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

isoleucine metabolic process
isoleucine catabolic process
leucine metabolic process
valine metabolic process
metabolic process
cellular amino acid biosynthetic process
branched-chain amino acid metabolic process
branched-chain amino acid biosynthetic process
branched-chain amino acid catabolic process
leucine biosynthetic process
regulation of hormone levels
catalytic activity
branched-chain-amino-acid transaminase activity
transaminase activity
transferase activity
L-leucine transaminase activity
L-valine transaminase activity
L-isoleucine transaminase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700007427.mm.1 NM_001243052.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007427.mm.1 NM_001243052.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007427.mm.1 TC0700000853.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700007431.mm.1 NM_001243053.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007431.mm.1 NM_001243053.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007431.mm.1 TC0700000853.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700007429.mm.1 NM_001243053.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007429.mm.1 NM_001243053.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007429.mm.1 TC0700000853.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700007421.mm.1 NM_009737.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007421.mm.1 NM_009737.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700007421.mm.1 TC0700000853.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700007434.mm.1 NM_001243052.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007434.mm.1 NM_001243052.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007434.mm.1 NM_001243053.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007434.mm.1 NM_001243053.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007434.mm.1 NM_009737.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007434.mm.1 NM_009737.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700007434.mm.1 TC0700000853.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700007435.mm.1 NM_001243052.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007435.mm.1 NM_001243052.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007435.mm.1 NM_001243053.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007435.mm.1 NM_001243053.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007435.mm.1 NM_009737.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007435.mm.1 NM_009737.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700007435.mm.1 TC0700000853.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 677903 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline