Assay Details
| Gene Symbol: Eif2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136070.1 | NP_001129542.1 | 11 - 12 | 1421 | 93 | |
| NM_010709.3 | NP_034839.2 | 11 - 12 | 1421 | 93 | ||
| GenBank mRNA | AK030312.1 | - | 11 - 12 | 1432 | 93 | |
| AK034115.1 | - | 11 - 12 | 1393 | 93 | ||
| AK036154.1 | - | 11 - 12 | 1417 | 93 | ||
| AK046539.1 | - | 11 - 12 | 1433 | 93 | ||
| AK047005.1 | - | 10 - 11 | 1266 | 93 | ||
| AK082748.1 | - | 11 - 12 | 1436 | 93 | ||
| AK083638.1 | - | 11 - 12 | 1381 | 93 | ||
| AK139052.1 | - | 11 - 12 | 1421 | 93 | ||
| AK161269.1 | - | 11 - 12 | 1404 | 93 | ||
| AK166085.1 | - | 11 - 12 | 1434 | 93 | ||
| AK167980.1 | - | 11 - 12 | 1417 | 93 | ||
| AK168120.1 | - | 11 - 12 | 1387 | 93 | ||
| AK170448.1 | - | 11 - 12 | 1433 | 93 | ||
| AK171115.1 | - | 11 - 12 | 1399 | 93 | ||
| BC023709.1 | - | 11 - 12 | 1374 | 93 | ||
| BC024501.1 | - | 6 - 7 | 735 | 93 | ||
| BC025036.1 | - | 11 - 12 | 1377 | 4935223 | 93 | |
| U58337.1 | - | 11 - 12 | 1306 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif2d
Genomic Map