This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00802914_m1
See other EIF2D GE Assays ›
Gene Symbol:
Eif2d
Gene Name:
eukaryotic translation initiation factor 2D
Gene Aliases:
D1Ertd5e, Lgtn
Chromosome Location:
Chr.1: 131153170 - 131187659 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00802914_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif2d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136070.1 NP_001129542.1 6 - 7 905 65
NM_010709.3 NP_034839.2 6 - 7 905 65
GenBank mRNA AK030312.1 - 6 - 7 917 65
AK034115.1 - 6 - 7 877 65
AK036154.1 - 6 - 7 901 65
AK046539.1 - 6 - 7 917 65
AK047005.1 - 6 - 7 900 65
AK082748.1 - 6 - 7 920 65
AK083638.1 - 6 - 7 865 65
AK139052.1 - 6 - 7 905 65
AK166085.1 - 6 - 7 918 65
AK167980.1 - 6 - 7 901 65
AK168120.1 - 6 - 7 871 65
AK170252.1 - 6 - 7 932 65
AK170448.1 - 6 - 7 917 65
AK171115.1 - 6 - 7 883 65
BC023709.1 - 6 - 7 858 65
BC024501.1 - 1 - 2 219 65
BC025036.1 - 6 - 7 861 4935223 65
U58337.1 - 6 - 7 790 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001168.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif2d

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
IRES-dependent translational initiation
translation
translational initiation
ribosome disassembly
RNA binding
translation initiation factor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100009398.mm.1 NM_001136070.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100009398.mm.1 NM_001136070.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100009398.mm.1 NM_010709.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100009398.mm.1 NM_010709.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100009398.mm.1 TC0100001168.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0