This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00803260_m1
See other RBL1 GE Assays ›
Gene Symbol:
Rbl1
Gene Name:
retinoblastoma-like 1 (p107)
Gene Aliases:
AW547426, PRB1, p107
Chromosome Location:
Chr.2: 157145893 - 157204534 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00803260_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001139516.1 NP_001132988.1 7 - 8 979 86
NM_011249.2 NP_035379.2 7 - 8 979 86
GenBank mRNA AK033402.1 - 7 - 8 973 86
AK147064.1 - 7 - 8 980 86
AK156055.1 - 7 - 8 958 86
AK178170.1 - 2 - 3 177 86
BC060124.1 - 7 - 8 946 6408844 86
U27177.1 - 7 - 8 963 86
U27178.1 - 7 - 8 963 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005015.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbl1

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle
chromatin modification
regulation of lipid kinase activity
regulation of cell cycle
protein binding
transcription factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200042924.mm.1 NM_001139516.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042924.mm.1 NM_001139516.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042924.mm.1 NM_011249.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042924.mm.1 NM_011249.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200042924.mm.1 TC0200005015.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0