This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00803381_g1
See other SARS GE Assays ›
Gene Symbol:
Sars
Gene Name:
seryl-aminoacyl-tRNA synthetase
Gene Aliases:
Sars1, Strs, serRS
Chromosome Location:
Chr.3: 108424864 - 108445259 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00803381_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sars
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204979.1 NP_001191908.1 8 - 9 1225 63
NM_011319.3 NP_035449.1 8 - 9 1225 63
GenBank mRNA AK028358.1 - 8 - 9 1157 63
AK075827.1 - 8 - 9 1156 63
AK077309.1 - 8 - 9 1106 63
AK082793.1 - 8 - 9 1136 63
AK152150.1 - 8 - 9 1180 63
AK153158.1 - 8 - 9 1175 63
BC008612.1 - 8 - 9 1114 3257101 63
BC031932.1 - 8 - 9 1115 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002742.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sars

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
tRNA aminoacylation for protein translation
seryl-tRNA aminoacylation
nucleotide binding
aminoacyl-tRNA ligase activity
serine-tRNA ligase activity
ATP binding
ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300021558.mm.1 NM_001204979.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021558.mm.1 NM_001204979.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021558.mm.1 NM_011319.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021558.mm.1 NM_011319.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300021558.mm.1 TC0300002742.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0