This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00803667_g1
See other IK GE Assays ›
Gene Symbol:
Ik
Gene Name:
IK cytokine
Gene Aliases:
MuRED
Chromosome Location:
Chr.18: 36744656 - 36757639 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00803667_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011879.2 NP_036009.1 12 - 13 1293 66
GenBank mRNA AJ006130.1 - 12 - 13 1176 66
AK082356.1 - 12 - 13 1225 66
AK087976.1 - 12 - 13 1239 66
BC006706.1 - 10 - 11 1651 66
BC014739.1 - 12 - 13 1224 4920184 66
BC026569.1 - 12 - 13 1224 4982612 66
BC029148.1 - 10 - 11 2187 66
BC029671.1 - 12 - 13 1236 5364953 66
BC029694.1 - 12 - 13 1233 5369477 66
BC033356.1 - 12 - 13 1220 5366230 66

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ik

Panther Classification:

Molecular Function -

cytokine

Gene Ontology Categories:

Process(es)

identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline