This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00803678_g1
See other IK GE Assays ›
Gene Symbol:
Ik
Gene Name:
IK cytokine
Gene Aliases:
MuRED
Chromosome Location:
Chr.18: 36744656 - 36757639 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00803678_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011879.2 NP_036009.1 5 - 6 521 79
GenBank mRNA AJ006130.1 - 5 - 6 404 79
AK029644.1 - 5 - 6 475 79
AK050625.1 - 5 - 6 479 79
AK082356.1 - 5 - 6 453 79
AK087976.1 - 5 - 6 467 79
AK088908.1 - 5 - 6 456 79
AK163766.1 - 5 - 6 475 79
AK167238.1 - 5 - 6 486 79
BC014739.1 - 5 - 6 452 4920184 79
BC026569.1 - 5 - 6 452 4982612 79
BC029148.1 - 5 - 6 494 79
BC029671.1 - 5 - 6 464 5364953 79
BC029694.1 - 5 - 6 461 5369477 79
BC033356.1 - 5 - 6 448 5366230 79
BG084120.2 - 5 - 6 521 79

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ik

Panther Classification:

Molecular Function -

cytokine

Gene Ontology Categories:

Process(es)

identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800002629.mm.1 NM_011879.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800002629.mm.1 NM_011879.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800002629.mm.1 TC1800000343.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800002627.mm.1 NM_011879.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800002627.mm.1 NM_011879.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800002627.mm.1 TC1800000343.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline