This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00803828_m1
See other TTPA GE Assays ›
Gene Symbol:
Ttpa
Gene Name:
tocopherol (alpha) transfer protein
Gene Aliases:
alpha-TTP
Chromosome Location:
Chr.4: 20008089 - 20030785 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00803828_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ttpa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015767.3 NP_056582.1 2 - 3 370 126
GenBank mRNA AF218416.1 - 2 - 3 358 126
AK004882.1 - 2 - 3 351 126
AK050374.1 - 2 - 3 371 126
BC026882.1 - 2 - 3 257 126
BC099868.1 - 2 - 3 369 40040166 126
BC099869.1 - 2 - 3 371 40040167 126
BC099870.1 - 2 - 3 371 40040169 126
BC099871.1 - 2 - 3 370 126

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000168.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ttpa

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

placenta development
embryonic placenta development
transport
response to pH
response to toxic substance
vitamin E metabolic process
intermembrane transport
vitamin transport
intracellular pH reduction
negative regulation of cell death
negative regulation of establishment of blood-brain barrier
transporter activity
phosphatidylinositol-4,5-bisphosphate binding
lipid binding
vitamin E binding
vitamin binding
phosphatidylinositol-3,4-bisphosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400001075.mm.1 NM_015767.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400001075.mm.1 TC0400000168.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline