This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00803829_m1
See other TTPA GE Assays ›
Gene Symbol:
Ttpa
Gene Name:
tocopherol (alpha) transfer protein
Gene Aliases:
alpha-TTP
Chromosome Location:
Chr.4: 20008089 - 20030785 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00803829_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer:
284.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ttpa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015767.3 NP_056582.1 3 - 4 560 76
GenBank mRNA AF218416.1 - 3 - 4 548 76
AK004882.1 - 3 - 4 541 76
AK050374.1 - 3 - 4 561 76
BC026882.1 - 3 - 4 447 76
BC099868.1 - 3 - 4 559 40040166 76
BC099869.1 - 3 - 4 561 40040167 76
BC099870.1 - 3 - 4 561 40040169 76
BC099871.1 - 3 - 4 560 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000168.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ttpa

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

placenta development
embryonic placenta development
transport
response to pH
response to toxic substance
vitamin E metabolic process
intermembrane transport
vitamin transport
intracellular pH reduction
negative regulation of cell death
negative regulation of establishment of blood-brain barrier
transporter activity
phosphatidylinositol-4,5-bisphosphate binding
lipid binding
vitamin E binding
vitamin binding
phosphatidylinositol-3,4-bisphosphate binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128446 AK050374.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128446 NM_015767.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013701 BC043705.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013701 BC099868.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013701 BC099869.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013701 BC099870.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013701 BC099871.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545011 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214190 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682480 AK016297.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682480 AK017883.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682480 AK086057.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 2502 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389839 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512052 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634067 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 934084 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 106493 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713763 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687222 AK015238.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687222 XM_977950.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687222 XM_980182.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 46768 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655438 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 42793 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159622 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8909 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 165525 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013701 BC043705.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013701 BC099868.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013701 BC099869.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013701 BC099870.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013701 BC099871.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128446 AK050374.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128446 NM_015767.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P29610 BC043705.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P29610 AK050374.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P29610 BC026882.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P29610 NM_015767.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P29610 AK004882.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P258817 AK050374.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767893 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767891 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767878 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767883 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767897 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767900 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767876 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767880 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767901 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767899 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767903 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767885 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767895 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767889 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767887 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767884 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767888 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767902 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22767881 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05172386 - -
Whole Mouse Genome G4122A A_51_P258817 AK050374.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P29610 BC043705.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P29610 AK050374.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P29610 BC026882.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P29610 NM_015767.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P29610 AK004882.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138481 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128446 AK050374.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128446 NM_015767.3 mouse_transcriptome
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767941 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767688 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767908 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767940 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767693 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767900 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767930 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767694 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767696 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767891 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767698 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767695 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767945 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767920 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767911 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767901 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767944 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767906 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767923 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767878 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767692 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767928 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767903 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767933 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767936 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767921 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767689 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767880 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767893 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767927 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767938 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767884 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767691 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767889 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767885 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767919 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767899 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767909 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767883 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767887 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767913 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767895 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767882 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22767925 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25632131 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767696 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767695 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767694 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767698 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P25632131 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10503504 - -
Affymetrix Mouse Gene 1.0 ST Array 10503521 - -
Affymetrix Mouse Gene 1.0 ST Array 10503503 - -
Affymetrix Mouse Gene 1.0 ST Array 10503506 - -
Affymetrix Mouse Gene 1.0 ST Array 10503505 - -
Affymetrix Mouse Gene 1.0 ST Array 10503507 - -
Affymetrix Mouse Gene 1.0 ST Array 10503522 - -
Affymetrix HT MG-430 PM Array Plate 1427284_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10503520 - -
Affymetrix Mouse Gene 1.1 ST Array 10503502 - -
Mouse Genome 430 2.0 Array 1427284_a_at - -
Mouse Genome 430 2.0 Array 1427284_PM_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 50500_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 50500_PM_at - -
Affymetrix Mouse Clariom S Array PSR0400001077.mm.1 NM_015767.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400001077.mm.1 TC0400000168.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400001076.mm.1 NM_015767.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400001076.mm.1 TC0400000168.mm.1 MTA TC
Mouse Expression Set 430 1427284_a_at - -
Mouse Expression Set 430 1427284_PM_a_at - -
Mouse Genome 430A 2.0 Array 1427284_a_at - -
Mouse Genome 430A 2.0 Array 1427284_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1427284_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1427284_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 819641 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 630576 - -
Illumina mouse-6 v1.0 expression beadchip 630576 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000630576 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229956 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1229956 - -
mouse-6 v1.1 (Illumina) scl25637.7_145-S - -
Illumina Sentrix mouse-6.v1 bead array 630576 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl25637.7_145-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 630576 - -
MouseRef-8 v2 Expression BeadChip ILMN_2470983 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2470983 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline