This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00804712_m1
See other SPINT4 GE Assays ›
Gene Symbol:
Spint4
Gene Name:
serine protease inhibitor, Kunitz type 4
Gene Aliases:
9230105I15Rik
Chromosome Location:
Chr.2: 164698501 - 164702448 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00804712_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spint4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030058.2 NP_084334.1 3 - 4 541 70
GenBank mRNA AK020315.1 - 3 - 4 514 70
AY226988.1 - 3 - 4 541 70
BC119299.1 - 3 - 4 511 8734048 70
BC119301.1 - 3 - 4 511 8734050 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002528.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Spint4

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
negative regulation of peptidase activity
molecular_function
serine-type endopeptidase inhibitor activity
peptidase inhibitor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492266 NM_030058.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492266 AK020315.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492266 AY226988.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492266 BC119299.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492266 BC119301.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492266 NM_030058.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492266 AK020315.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492266 AY226988.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492266 BC119299.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492266 BC119301.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394212 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920474 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11450 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118125 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492266 NM_030058.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492266 AK020315.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492266 AY226988.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492266 BC119299.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492266 BC119301.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492266 NM_030058.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492266 AK020315.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492266 AY226988.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492266 BC119299.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492266 BC119301.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819276 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819257 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819260 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819268 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819286 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819263 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819261 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819259 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819264 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819265 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819266 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819283 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819272 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819280 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819282 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819269 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819277 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819270 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819278 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819273 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819285 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118125 - -
Whole Mouse Genome G4122A A_51_P492266 NM_030058.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P492266 AK020315.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P492266 AY226988.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P492266 BC119299.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P492266 BC119301.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P492266 NM_030058.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P492266 AK020315.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P492266 AY226988.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P492266 BC119299.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P492266 BC119301.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114967 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120512 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118125 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01705674 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01705670 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02070765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819268 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819270 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819276 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819300 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819277 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819258 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819285 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819251 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819263 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819328 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819252 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819272 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819266 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819265 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819322 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819239 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819303 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819289 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819302 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819280 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819249 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819299 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819254 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819297 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819269 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819230 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819295 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819327 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819282 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819247 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819227 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819257 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819318 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819259 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819245 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819330 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01755522 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819234 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819268 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819260 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819305 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819289 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02040564 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819249 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819290 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819278 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819251 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819297 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819270 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819263 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819291 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819272 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819266 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819277 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01525583 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819261 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819239 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819282 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02237555 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819227 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819269 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819293 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819241 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819230 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819229 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819247 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02221560 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819280 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819233 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819295 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01156366 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819308 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819302 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819252 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819285 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819258 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819299 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01955643 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819300 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01745355 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819276 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819307 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01705673 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819274 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819264 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819259 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819287 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819265 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819257 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10478550 - -
Affymetrix Mouse Gene 1.0 ST Array 10478548 - -
Affymetrix Mouse Gene 1.0 ST Array 10478552 - -
Affymetrix Mouse Gene 1.0 ST Array 10478553 - -
Affymetrix Mouse Gene 1.0 ST Array 10478549 - -
Affymetrix Mouse Gene 1.0 ST Array 10478551 - -
Affymetrix HT MG-430 PM Array Plate 1431444_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10478547 - -
Mouse Genome 430 2.0 Array 1431444_at - -
Mouse Genome 430 2.0 Array 1431444_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 78239_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 78239_PM_at - -
Affymetrix Mouse Clariom S Array PSR0200020649.mm.1 NM_030058.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020649.mm.1 NM_030058.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200020649.mm.1 TC0200002528.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200020647.mm.1 NM_030058.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020647.mm.1 NM_030058.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200020647.mm.1 TC0200002528.mm.1 MTA TC
Mouse Expression Set 430 1431444_PM_at - -
Mouse Expression Set 430 1431444_at - -
Affymetrix HT Mouse Genome 430B Array 1431444_at - -
Affymetrix HT Mouse Genome 430B Array 1431444_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 339027 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 840577 - -
Illumina mouse-6 v1.0 expression beadchip 840577 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000840577 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240354 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1240354 - -
mouse-6 v1.1 (Illumina) scl19929.4.1_6-S - -
Illumina Sentrix mouse-6.v1 bead array 840577 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl19929.4.1_6-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 840577 - -
MouseRef-8 v2 Expression BeadChip ILMN_1240354 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240354 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline