This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00805235_m1
See other HADHA GE Assays ›
Gene Symbol:
Hadha
Gene Name:
hydroxyacyl-Coenzyme A dehydrogenase/3-ketoacyl-Coenzyme A thiolase/enoyl-Coenzyme A hydratase (trifunctional protein), alpha subunit
Gene Aliases:
C77020, Mtpa
Chromosome Location:
Chr.5: 30118304 - 30154980 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00805235_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hadha
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178878.2 NP_849209.1 3 - 4 211 103
GenBank mRNA AK029017.1 - 3 - 4 212 103
AK035399.1 - 3 - 4 194 103
AK053986.1 - 3 - 4 209 103
AK150188.1 - 3 - 4 209 103
AK170478.1 - 3 - 4 212 103
AK170683.1 - 3 - 4 212 103
AK171469.1 - 3 - 4 212 103
AK218789.1 - 2 - 3 88 103
BC037009.1 - 3 - 4 199 5134605 103
BC046978.1 - 3 - 4 185 6315204 103
BC058569.1 - 3 - 4 207 6468514 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002154.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hadha

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
fatty acid beta-oxidation
metabolic process
response to insulin
response to drug
oxidation-reduction process
fatty-acyl-CoA binding
catalytic activity
3-hydroxyacyl-CoA dehydrogenase activity
acetyl-CoA C-acyltransferase activity
enoyl-CoA hydratase activity
oxidoreductase activity
long-chain-enoyl-CoA hydratase activity
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
lyase activity
protein complex binding
NAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500019538.mm.1 TC0500002154.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500019538.mm.1 NM_178878.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019538.mm.1 NM_178878.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wxzsf:80/100.66.72.186:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0