This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00805372_g1
See other GPT GE Assays ›
Gene Symbol:
Gpt
Gene Name:
glutamic pyruvic transaminase, soluble
Gene Aliases:
1300007J06Rik, 2310022B03Rik, ALT, Gpt-1, Gpt1
Chromosome Location:
Chr.15: 76696764 - 76699675 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00805372_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182805.2 NP_877957.1 10 - 11 1502 84
GenBank mRNA AK008086.1 - 10 - 11 1526 84
AK009457.1 - 10 - 11 1525 84
BC022625.1 - 10 - 11 1503 4195300 84
BC026846.1 - 10 - 11 1501 4194336 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000605.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpt

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

biosynthetic process
catalytic activity
L-alanine:2-oxoglutarate aminotransferase activity
transaminase activity
transferase activity
pyridoxal phosphate binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350022 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426739 AK009457.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426739 BC022625.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426739 BC026846.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426739 NM_182805.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426739 AK008086.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426739 AK009457.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426739 BC022625.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426739 BC026846.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426739 NM_182805.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426739 AK008086.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458500 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134869 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426739 AK009457.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426739 BC022625.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426739 BC026846.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426739 NM_182805.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426739 AK008086.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426739 AK009457.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426739 BC022625.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426739 BC026846.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426739 NM_182805.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426739 AK008086.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426739 AK009457.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426739 BC022625.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426739 BC026846.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426739 NM_182805.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426739 AK008086.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426739 AK009457.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426739 BC022625.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426739 BC026846.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426739 NM_182805.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426739 AK008086.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350010 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350023 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350002 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350005 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350026 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30349998 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350000 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350031 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350001 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350034 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350025 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350007 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350035 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350013 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350012 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350004 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350033 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350016 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350029 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30349995 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350014 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350003 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350006 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350018 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350024 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350022 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350020 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30349996 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134869 - -
Whole Mouse Genome G4122A A_51_P426739 AK009457.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426739 BC022625.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426739 BC026846.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426739 NM_182805.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426739 AK008086.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426739 AK009457.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426739 BC022625.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426739 BC026846.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426739 NM_182805.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426739 AK008086.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102303 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118269 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134869 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12362724 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12362741 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12362782 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12362792 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12362690 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12362756 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350022 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350029 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350089 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350096 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350031 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350072 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350007 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350081 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350075 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350113 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350112 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350004 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350068 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350097 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30349996 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350053 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350022 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350091 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350088 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350082 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350002 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350094 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350107 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350050 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350058 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30349998 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350061 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350038 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350048 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350024 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350100 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350012 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350111 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350034 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350035 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350092 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350016 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350025 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350055 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350042 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350020 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350026 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350040 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350070 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350118 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350064 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350033 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350114 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350023 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350046 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350084 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350018 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350014 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350105 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350000 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350006 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350071 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350010 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350104 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12362756 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350006 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30349996 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350002 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350029 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350034 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12362739 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350012 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350031 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350010 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12362697 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12362828 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350020 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350025 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350004 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12362822 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350000 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350035 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12362800 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350022 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350013 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350018 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30350016 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10424984 - -
Affymetrix Mouse Gene 1.0 ST Array 10424983 - -
Affymetrix Mouse Gene 1.0 ST Array 10424990 - -
Affymetrix Mouse Gene 1.0 ST Array 10424985 - -
Affymetrix Mouse Gene 1.0 ST Array 10424986 - -
Affymetrix Mouse Gene 1.0 ST Array 10424989 - -
Affymetrix Mouse Gene 1.0 ST Array 10424980 - -
Affymetrix Mouse Gene 1.0 ST Array 10424988 - -
Affymetrix Mouse Gene 1.0 ST Array 10424982 - -
Affymetrix Mouse Gene 1.0 ST Array 10424981 - -
Affymetrix Mouse Gene 1.0 ST Array 10424987 - -
Affymetrix HT MG-430 PM Array Plate 1426502_PM_s_at - -
Affymetrix HT MG-430 PM Array Plate 1453216_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10424979 - -
Mouse Genome 430 2.0 Array 1426502_s_at - -
Mouse Genome 430 2.0 Array 1426502_PM_s_at - -
Mouse Genome 430 2.0 Array 1453216_PM_at - -
Mouse Genome 430 2.0 Array 1453216_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 76282_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 76282_at - -
Affymetrix Mouse Clariom S Array PSR1500004421.mm.1 NM_182805.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500004421.mm.1 TC1500000605.mm.1 MTA TC
Mouse Expression Set 430 1426502_PM_s_at - -
Mouse Expression Set 430 1426502_s_at - -
Mouse Expression Set 430 1453216_PM_at - -
Mouse Expression Set 430 1453216_at - -
Murine Genome U74v2 Set 132388_f_at - -
Murine Genome U74v2 Set 137237_PM_at - -
Murine Genome U74v2 Set 137237_at - -
Murine Genome U74v2 Set 132388_PM_f_at - -
Mouse Genome 430A 2.0 Array 1426502_s_at - -
Mouse Genome 430A 2.0 Array 1426502_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1426502_s_at - -
Affymetrix HT Mouse Genome 430A Array 1426502_PM_s_at - -
Affymetrix HT Mouse Genome 430B Array 1453216_at - -
Affymetrix HT Mouse Genome 430B Array 1453216_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 2350270 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002350270 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2683924 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2683924 - -
mouse-6 v1.1 (Illumina) scl47787.2_58-S - -
Illumina Sentrix mouse-6.v1 bead array 2350270 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl47787.2_58-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2350270 - -
MouseRef-8 v2 Expression BeadChip ILMN_2832524 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2832524 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl47787.2_58-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0