This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00805380_g1
See other GPT GE Assays ›
Gene Symbol:
Gpt
Gene Name:
glutamic pyruvic transaminase, soluble
Gene Aliases:
1300007J06Rik, 2310022B03Rik, ALT, Gpt-1, Gpt1
Chromosome Location:
Chr.15: 76696764 - 76699675 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00805380_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182805.2 NP_877957.1 8 - 9 1235 64
GenBank mRNA AK008086.1 - 8 - 9 1258 64
AK009457.1 - 8 - 9 1258 64
BC022625.1 - 8 - 9 1236 4195300 64
BC026846.1 - 8 - 9 1234 4194336 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000605.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpt

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

biosynthetic process
catalytic activity
L-alanine:2-oxoglutarate aminotransferase activity
transaminase activity
transferase activity
pyridoxal phosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500004417.mm.1 NM_182805.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500004417.mm.1 TC1500000605.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0