This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00805459_g1
See other ASIC3 GE Assays ›
Gene Symbol:
Asic3
Gene Name:
acid-sensing (proton-gated) ion channel 3
Gene Aliases:
AW742291, Accn3, DRASIC, SLNAC1, TNAC1
Chromosome Location:
Chr.5: 24410289 - 24417834 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00805459_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Asic3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183000.2 NP_892045.2 10 - 11 1778 88
GenBank mRNA AY261387.1 - 9 - 10 1369 88
BC054460.1 - 9 - 10 2047 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000214.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Ion Channel 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Asic3

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
sodium ion transport
response to heat
response to mechanical stimulus
response to acidic pH
ion transmembrane transport
sodium ion transmembrane transport
enterobactin transport
detection of chemical stimulus involved in sensory perception
sensory perception of sour taste
detection of temperature stimulus involved in sensory perception
detection of temperature stimulus involved in sensory perception of pain
detection of mechanical stimulus involved in sensory perception of pain
detection of chemical stimulus involved in sensory perception of pain
detection of mechanical stimulus involved in sensory perception
cation transmembrane transport
cation channel activity
sodium channel activity
ligand-gated sodium channel activity
PDZ domain binding
enterobactin transporter activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162727 BC054460.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162727 NM_183000.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132496 AY261387.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132496 AK028403.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132496 BC054460.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132496 NM_183000.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580551 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663714 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143427 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154505 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153308 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121036 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162727 BC054460.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162727 NM_183000.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P609738 AY261387.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P609738 BC054460.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P609738 NM_183000.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P609738 AK028403.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541649 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541662 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541639 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541636 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541657 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541642 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541644 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541653 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541646 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541651 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541625 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541634 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541624 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541655 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541633 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541661 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541643 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541637 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541660 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541628 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541630 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541622 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541663 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541626 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541647 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121036 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153308 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154505 - -
Whole Mouse Genome G4122A A_52_P609738 AY261387.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P609738 BC054460.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P609738 NM_183000.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P609738 AK028403.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129195 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132496 AY261387.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132496 AK028403.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132496 BC054460.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132496 NM_183000.2 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154505 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121036 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153308 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04675100 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04675092 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04675086 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04675087 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04675084 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04675105 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541672 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541644 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541724 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541727 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541754 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541701 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541695 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541675 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541667 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541745 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541702 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541696 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541668 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541649 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541714 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541756 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541720 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541679 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541717 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541729 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541726 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541739 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541728 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541674 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541633 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541637 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541742 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541734 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541623 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541737 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541625 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541716 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541751 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541630 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541741 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541749 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541686 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541722 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541676 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541700 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541747 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541655 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541665 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541670 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541639 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541651 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541718 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541752 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541708 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541680 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541709 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541731 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541661 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541634 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541697 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541688 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541735 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541671 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541677 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541711 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04771967 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04771950 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541667 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04462405 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04771958 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04835456 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04392530 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541671 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04675104 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541670 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541651 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541649 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541676 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04835457 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04392531 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04750053 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04675092 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541647 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541668 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04771957 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04771956 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541662 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04771965 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541638 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541634 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541641 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541625 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04771955 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04675087 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541677 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541646 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04873907 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541674 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541630 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541655 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541675 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541644 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04835432 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541672 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23541679 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10520180 - -
Affymetrix Mouse Gene 1.0 ST Array 10520184 - -
Affymetrix Mouse Gene 1.0 ST Array 10520179 - -
Affymetrix Mouse Gene 1.0 ST Array 10520174 - -
Affymetrix Mouse Gene 1.0 ST Array 10520178 - -
Affymetrix Mouse Gene 1.0 ST Array 10520183 - -
Affymetrix Mouse Gene 1.0 ST Array 10520176 - -
Affymetrix Mouse Gene 1.0 ST Array 10520181 - -
Affymetrix Mouse Gene 1.0 ST Array 10520186 - -
Affymetrix Mouse Gene 1.0 ST Array 10520182 - -
Affymetrix Mouse Gene 1.0 ST Array 10520185 - -
Affymetrix Mouse Gene 1.0 ST Array 10520175 - -
Affymetrix HT MG-430 PM Array Plate 1439983_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10520173 - -
Mouse Genome 430 2.0 Array 1439983_PM_a_at - -
Mouse Genome 430 2.0 Array 1439983_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 171209_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 171209_PM_at - -
Affymetrix Mouse Clariom S Array PSR0500001594.mm.1 TC0500000214.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500001594.mm.1 NM_183000.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500001594.mm.1 NM_183000.2 Refseq
Mouse Expression Set 430 1439983_PM_a_at - -
Mouse Expression Set 430 1439983_a_at - -
Affymetrix HT Mouse Genome 430B Array 1439983_PM_a_at - -
Affymetrix HT Mouse Genome 430B Array 1439983_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 316711 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 6940672 - -
Illumina mouse-6 v1.0 expression beadchip 4200524 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006940672 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004200524 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2699096 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2615124 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2699096 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2615124 - -
mouse-6 v1.1 (Illumina) scl0004047.1_78-S - -
mouse-6 v1.1 (Illumina) scl0004191.1_1-S - -
Illumina Sentrix mouse-6.v1 bead array 6940672 - -
Illumina Sentrix mouse-6.v1 bead array 4200524 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0004047.1_78-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0004191.1_1-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4200524 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940672 - -
MouseRef-8 v2 Expression BeadChip ILMN_2830611 - -
MouseRef-8 v2 Expression BeadChip ILMN_2699096 - -
MouseRef-8 v2 Expression BeadChip ILMN_2615124 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2830611 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2615124 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2699096 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0