This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00805557_g1
See other ASIC4 GE Assays ›
Gene Symbol:
Asic4
Gene Name:
acid-sensing (proton-gated) ion channel family member 4
Gene Aliases:
Accn4, BNAC4
Chromosome Location:
Chr.1: 75450510 - 75474340 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00805557_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Asic4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183022.3 NP_898843.1 8 - 9 1720 66
GenBank mRNA BC046481.1 - 8 - 9 1353 66
BC055772.1 - 8 - 9 1531 6808924 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000660.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Asic4

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
sodium ion transport
sodium channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100005668.mm.1 NM_183022.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005668.mm.1 NM_183022.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100005668.mm.1 TC0100000660.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0