This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00805725_m1
See other SPINKL GE Assays ›
Gene Symbol:
Spinkl
Gene Name:
serine protease inhibitor, Kazal type-like
Gene Aliases:
9530002K18Rik
Chromosome Location:
Chr.18: 44166358 - 44176707 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00805725_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spinkl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183123.2 NP_898946.1 2 - 3 113 151
GenBank mRNA AK020535.1 - 2 - 3 114 151

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK020535.1, NM_183123.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Spinkl

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s)

positive regulation of sperm motility
negative regulation of sperm capacitation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800009524.mm.1 NM_183123.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800009524.mm.1 NM_183123.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800009524.mm.1 TC1800001278.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline