This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00812562_m1
See other ARHGAP44 GE Assays ›
Gene Symbol:
Arhgap44
Gene Name:
Rho GTPase activating protein 44
Gene Aliases:
6330543G20, AI840762, AU040829, AW493732, Rich2
Chromosome Location:
Chr.11: 65002039 - 65163120 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00812562_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap44
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001099288.1 NP_001092758.1 4 - 5 598 100
NM_175003.3 NP_778168.2 4 - 5 598 100
GenBank mRNA AK053220.1 - 4 - 5 326 100
BC056366.1 - 4 - 5 598 6825221 100
BC059911.1 - 4 - 5 598 6825221 100

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap44

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
GTPase activator activity
phospholipid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0